Learn More
Kartell™ Faeces Container
Easy means of collecting and transporting faeces samples. Kartell™ Faeces Container includes a spoon to assist in handling the sample.
Brand:  Kartell™ 0259500
Product Code. 10031081
  • 1545.00 SEK / Pack of 360
Description
  • Particularly designed for collection and delivery of faeces
  • Made of transparent PS with stopper of PP
  • Incorporates a small spoon to facilitate sample handling
  • Sterile
  • CE Marked and IVD Certified
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Faeces Container > CapacityMetric: 60mL