Learn More
Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device Product Code.: 12005635
Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device
Product Code. 12005635
50 kDa, White, 44 mm, Pack of 12
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
20 kDa
3.5 to 5 kDa
50 kDa
8 to 10 kDa
Closure Color:
Black
Blue
Green
Orange
Red
White
Yellow
Height (Metric):
44 mm
63 mm
Unit Size:
Pack of 12
Product Code. 12005635
Brand: Spectrum™ F235058
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever
Ready-to-Use and disposable, the Float-A-Lyzer G2 incorporates Biotech Grade Cellulose Ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use & provides ultimate convenience. Available in volume sizes: 1, 5, and 10 mL. The resealable screw cap is color-coded for 9 available MWCO's and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5 and 10mL sizes). The flotation ring (included) provides buoyancy to maximize dialysis efficiency.
  • Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
  • Leak-proof Luer-Lok™ cap provides easy access with the included syringe for easy loading and sample retrieval
  • Units can also interlock for the simultaneous dialysis of multiple samples
  • Ready-to-use and complete
  • Easy syringe loading (no needle)
  • Total sample recovery
  • Ultrapure Biotech CE (no cleaning)
  • Largest range of MWCOs
  • Maximizes SA & minimizes hold-up volume
Specifications
Product Type Micro Float-A-Lyzer Dialysis Device
MWCO 50 kDa
Membrane Cellulose Ester
Material Polycarbonate
Diameter (English) Membrane 0.25 in.
Diameter (Metric) Membrane 6.4 mm
Height (English) 1.7 in.
Height (Metric) 44 mm
Width (English) 1.77 in.
Width (Metric) 45 mm
Capacity (Metric) 100 to 200 ÎĽL
Closure Color White
Closure Material Polypropylene
Closure Type Luer lok
Kit Contents 1mL syringe
Quantity Each
For Use With (Application) Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
Show More Show Less
Product Content Correction
Product Title
Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device >