Learn More
Nabertherm™ Split Type Vertical Tube Furnace RSV 1300 Series with B500 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
74316.01 SEK - 167329.36 SEK
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17358119
|
Nabertherm™
RI152K3AN |
50 mm | 250 mm |
74316.01 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17378119
|
Nabertherm™
RI252K3AN |
50 mm | 500 mm |
82599.64 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17318129
|
Nabertherm™
RI322K3AN |
120 mm | 500 mm |
104610.43 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17358129
|
Nabertherm™
RI382K3AN |
80 mm | 500 mm |
101533.66 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17398129
|
Nabertherm™
RI422K3AN |
120 mm | 750 mm |
135023.19 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17338139
|
Nabertherm™
RI472K3AN |
170 mm | 750 mm |
149223.71 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17378139
|
Nabertherm™
RI482K3AN |
80 mm | 750 mm |
129579.66 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17318149
|
Nabertherm™
RI522K3AN |
120 mm | 1000 mm |
147921.99 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17358149
|
Nabertherm™
RI572K3AN |
170 mm | 1000 mm |
167329.36 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1300°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type S thermocouple
- Frame for vertical operation
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 includes fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- RSV: switchgear and control unit separate from furnace in own wall or standing cabinet
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
B500 | |
Up to 1300°C |