Learn More
Nabertherm™ Split Type Vertical Tube Furnace RSV 1300 Series with P570 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
98456.88 SEK - 183186.59 SEK
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17398119
|
Nabertherm™
RI252V3AN3 |
50 mm | 500 mm |
98456.88 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17338129
|
Nabertherm™
RI322V3AN3 |
120 mm | 500 mm |
121651.05 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17378129
|
Nabertherm™
RI382V3AN3 |
80 mm | 500 mm |
117390.89 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17318139
|
Nabertherm™
RI422V3AN3 |
120 mm | 750 mm |
150880.43 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17358139
|
Nabertherm™
RI472V3AN3 |
170 mm | 750 mm |
165080.94 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17398139
|
Nabertherm™
RI482V3AN3 |
80 mm | 750 mm |
145436.90 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17338149
|
Nabertherm™
RI522V3AN3 |
120 mm | 1000 mm |
163779.23 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
17378149
|
Nabertherm™
RI572V3AN3 |
170 mm | 1000 mm |
183186.59 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1300°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type S thermocouple
- Frame for vertical operation
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 includes fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- RSV: switchgear and control unit separate from furnace in own wall or standing cabinet
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
P570 | |
Up to 1300°C |