Learn More
GVS SPR582 Pair of ABE1P3 Replacement Filters for Elipse Masks
These ABE1P3 filters are recommended for multigas and dust risks (excluding ammonia), in chemical production and handling environment. Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, and dust.
Brand: GVS SPR582
Description
These ABE1P3 filters are recommended for multigas and dust risks (excluding ammonia), in chemical production and handling environment. Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, and dust.
- These filters are compatible with our ABE1P3 masks, SPR580 (S/M) and SPR581 (M/L).Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
- Filters last up to 3 years and can be easily replaced.
- Gas Filters: Activated carbon sealed into a ABS shell.
- Particulate filters: Mechanical type multi-layer HESPA Synthetic media with TPE flexible overmolded/encapsulated.
- Particulate filters are integrated with the carbon element.
- Certifications: ABE1P3 R D Filters EN14387: 2004 + A1: 2008.
Specifications
Gray | |
Filter | |
No |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.