missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRR1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11959-1-AP-20UL
This item is not returnable.
View return policy
Description
SPRR1B is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. All that results in the formation of an insoluble envelope beneath the plasma membrane. Can function as both amine donor and acceptor in transglutaminase-mediated cross-linkage.Specifications
SPRR1B | |
Polyclonal | |
Unconjugated | |
SPRR1B | |
14.9 kDa pancornulin, CORNIFIN, Cornifin B, GADD33, Small proline rich protein IB, SPR IB, SPRR1, SPRR1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6699 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P22528 | |
SPRR1B | |
SPRR1B Fusion Protein Ag2563 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SPRR1B Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction