missing translation for 'onlineSavingsMsg'
Learn More

SPRR2F Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24648-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883135

  • 1355.00 SEK / 20µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cross-linked envelope protein of keratinocytes. It is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. All that results in the formation of an insoluble envelope beneath the plasma membrane.
Specifications

Specifications

SPRR2F
Polyclonal
Unconjugated
SPRR2F
small proline rich protein 2F, SPR 2F, SPRR2F
Rabbit
Antigen Affinity Chromatography
RUO
6705
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.52 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96RM1
SPRR2F
SPRR2F Fusion Protein Ag20323
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers