Läs mer
SPTLC1 Mouse anti-Human, Unconjugated, Clone: 2D11D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66899-1-IG-150UL
Beskrivning
The SPTLC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SPTLC1. This antibody recognizes human, mouse, rat antigen. The SPTLC1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifikationer
| SPTLC1 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 2D11D7 | |
| Unconjugated | |
| 150 μL | |
| 10558 | |
| IgG1 |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.