Learn More
SRC Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11097-1-AP-20UL
 View more versions of this product
Product Code. 16887943
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
SRC is a non-receptor kinase involved in the regulation of growth and differentiation of eukaryotic cells. SRC associates with the cytoplasmic region of various cell surface receptors to facilitate signal transduction, and is also involved in cytoskeletal organization, cell cycle regulation, and apoptosis. In humans, the gene encoding SRC is present on chromosome 20. Activity of SRC is regulated by two tyrosine residues (Tyr 416 and Tyr527), and phosphorylation of these two sites have opposing effects. Phosphorylation of Tyr416 in the activation loop of the kinase domain upregulates enzyme activity while the phosphorylation of Tyr527 decreases enzyme activity. Mutations in the SRC gene could be involved in the maligt progression of colon cancer. Two transcript variants encoding the same SRC protein have been found thus far.
Specifications
Specifications
SRC
Polyclonal
Unconjugated
SRC
ASV, c SRC, p60 Src, pp60c src, Proto oncogene c Src, SRC, SRC1
Rabbit
Antigen Affinity Chromatography
RUO
100757188, 20779, 6714, 83805
-20°C
Liquid
Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05480, P12931, Q9WUD9
SRC
SRC Fusion Protein Ag1575
20 ÎĽL
Primary
Human, Rat, Mouse, Hamster
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SRC Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated