Learn More
SRD5A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26001-1-AP-20UL
 View more versions of this product
Product Code. 16884525
  • 1645.54 SEK / 20µL
Description
Steroid 5alpha-Reductase is an important enzyme in androgen physiology because it catalyzes the conversion of testosterone into the more potent 5alpha-dihydro-testosterone, which mediates androgen effects on target tissues. The enzyme exists as two isoforms: type 1, which is expressed mainly in the skin; and type 2, which is expressed mainly in the prostate. In cultured human skin cells, 5alpha-Reductase 1 shows heterogeneity of protein, and has different levels of transcriptional and translational expression. 5alpha-Reductase 1 is expressed in all portions of the hair follicle, whereas 5alpha-Reductase 2 is expressed only in mesenchymal portions. In addition, 5alpha-Reductase 1 is mainly expressed in human breast carcinoma and may play a role in the in situ production and actions of the potent androgen 5alpha-dihydrotestosterone, including inhibition of cancer cell proliferation in hormone-dependent human breast carcinoma. The 5alpha-Reductase-3alpha-hydroxysteroid dehydrogenase complex is present in the human brain, suggesting that the complex may be involved in the synthesis of neuroactive steroids or the catabolism of neurotoxic steroids.
Specifications
Specifications
SRD5A1
Polyclonal
Unconjugated
SRD5A1
S5AR 1, SR type 1, SRD5A1, Steroid 5 alpha reductase 1
Rabbit
Antigen Affinity Chromatography
RUO
6715, 78925
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P18405, Q68FF9
SRD5A1
SRD5A1 Fusion Protein Ag23351
20 μL
Primary
Mouse, Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SRD5A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated