missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12090-1-AP-150UL
This item is not returnable.
View return policy
Description
The gene encodes a subunit of the endoplasmic reticulum signal recognition particle receptor that, in conjunction with the signal recognition particle, is involved in the targeting and translocation of signal sequence tagged secretory and membrane proteins across the endoplasmic reticulum. Alternative splicing results in multiple transcript variants.Specifications
SRPR | |
Polyclonal | |
Unconjugated | |
SRPRA | |
Docking protein alpha, DP, DP alpha, SR alpha, Sralpha, SRP alpha, SRPR, SRPR alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315548, 6734, 67398 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08240, Q9DBG7 | |
Srpr, Srpra | |
SRPR Fusion Protein Ag2730 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SRPR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction