Läs mer
SSEA-1 Antibody (4E10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-22179-0.025ml
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SSEA-1 Monoclonal specifically detects SSEA-1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifikationer
| CD15/Lewis X | |
| Monoclonal | |
| Unconjugated | |
| CD 15, CD-15, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| Mouse | |
| 59 kDa | |
| 0.025 mL | |
| Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers | |
| 2526 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 40000000000 | |
| ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| FUT4 | |
| Synthesized peptide of human CD15. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering