Learn More
SSRP1 Mouse anti-Human, Unconjugated, Clone: 3D3H4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67313-1-IG-150UL
Description
The SSRP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SSRP1. This antibody recognizes human, mouse, rat antigen. The SSRP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SSRP1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
3D3H4 | |
Unconjugated | |
150 μL | |
6749 | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.