Learn More
SSRP1 Mouse anti-Human, Unconjugated, Clone: 3D3H4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67313-1-IG-20UL
 View more versions of this product
Product Code. 16889865
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The SSRP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SSRP1. This antibody recognizes human, mouse, rat antigen. The SSRP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SSRP1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3D3H4
Unconjugated
20 μL
6749
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SSRP1 Mouse anti-Human, Unconjugated, Clone: 3D3H4, Proteintech > 20 μL