Learn More
SSRP1 Mouse anti-Human, Unconjugated, Clone: 3D3H4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67313-1-IG-150UL
 View more versions of this product
Product Code. 16879865
  • 4652.90 SEK / 150µL
Description
The SSRP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SSRP1. This antibody recognizes human, mouse, rat antigen. The SSRP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SSRP1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3D3H4
Unconjugated
150 μL
6749
IgG2b
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SSRP1 Mouse anti-Human, Unconjugated, Clone: 3D3H4, Proteintech > 150 μL