missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSTR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20587-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Somatostatin acts to regulate numerous physiological processes by binding to and activating specific receptors in target tissues. Activation of these receptors by somatostatin, which is secreted by nerve and endocrine cells, regulates the secretion of insulin, glucagon and growth hormone, neuronal excitability in both the brain and the peripheral nervous system, and cell growth. Somatostatin receptors have been implicated in numerous diseases ranging from Alzheimer′s to cancers of the gastrointestinal tract, breast, prostate, and pituitary.Specifications
SSTR1 | |
Polyclonal | |
Unconjugated | |
SSTR1 | |
somatostatin receptor 1, Somatostatin receptor type 1, SRIF 2, SS 1 R, SS1 R, SS1R, SSTR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20605, 25033, 6751 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P28646, P30872, P30873 | |
SSTR1 | |
SSTR1 Fusion Protein Ag14287 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |