Learn More
SSTR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20404-1-AP-150UL
This item is not returnable.
View return policy
Description
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.Specifications
SSTR2 | |
Polyclonal | |
Unconjugated | |
SSTR2 | |
somatostatin receptor 2, Somatostatin receptor type 2, SRIF 1, SS 2 R, SS2 R, SS2R, SSTR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6752 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P30874 | |
SSTR2 | |
SSTR2 Fusion Protein Ag14205 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
SSTR2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction