Learn More
SSTR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20404-1-AP-150UL
 View more versions of this product
Product Code. 16889934
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
Specifications
Specifications
SSTR2
Polyclonal
Unconjugated
SSTR2
somatostatin receptor 2, Somatostatin receptor type 2, SRIF 1, SS 2 R, SS2 R, SS2R, SSTR2
Rabbit
Antigen Affinity Chromatography
RUO
6752
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P30874
SSTR2
SSTR2 Fusion Protein Ag14205
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SSTR2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated