missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3GAL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24925-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
ST3GAL3 | |
Polyclonal | |
Unconjugated | |
ST3GAL3 | |
Alpha 2,3 ST 3, Sialyltransferase 6, SIAT6, ST3Gal III, ST3GAL3, ST3GALII, ST3GalIII, ST3N | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20441, 6487 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q11203 | |
ST3GAL3 | |
ST3GAL3 Fusion Protein Ag20795 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ST3GAL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction