missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stabilin-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Stabilin-1 |
---|---|
Concentration | 0.9 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890235
|
Proteintech
20663-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880235
|
Proteintech
20663-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 fasciclin, 16 epidermal growth factor (EGF)-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluro-binding Link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node.Specifications
Stabilin-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9NY15 | |
23166 | |
Peptide | |
Primary | |
-20°C | |
STAB1 |
0.9 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CLEVER 1, FEEL 1, FEEL1, FELE 1, FEX1, KIAA0246, MS 1 antigen, STAB 1, STAB1, Stabilin-1 | |
STAB1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Stabilin-1 Rabbit anti-Human, Polyclonal, Proteintech