Learn More
Bochem™ Cork Bore Set with Ejector
Brand: Bochem™ 12433
Additional Details : Weight : 0.80000kg
Specifications
Cork Borer Set | |
Tube: Brass, nickel plated; Cork borer: Brass chromed; Ejector: Stainless steel w/ plastic knob | |
12Pack |
Cork Borer Set | |
Sizes 16.25, 17.5, 18.75 |