Learn More
Trajan Scientific and Medical™ Stainless-Steel GC Unions
Provide low dead volume connections for optimum chromatography
Brand:  Trajan Scientific and Medical™ 103330
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15269103
  • 740.00 SEK / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Low dead volume
  • For high temperature applications
  • Leak-free connections
  • Does not require back ferrules
  • Used with Graphite, Graphite Vespel™ or PTFE ferrules
  • Suitable for glass, fused silica, metal, plastic or glass lined tubing
  • Non-swaging and no tube distortion
Specifications
Specifications
Union
0.0625 x 0.0625 in.