Learn More
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW351-02
5538.09 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10104131
  • 5660.00 SEK / Each
Only null left
Estimated Shipment: 23-07-2025
Specifications
Wood
5 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight With DAkkS Calibration Certificate > Mass: 5g