Learn More
Sartorius™ Stainless Steel OIML Class F1 Calibration Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW4238-02
1804.42 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10738002
  • 1924.00 SEK / Each
Only null left
Estimated Shipment: 27-08-2025
Specifications
Plastic
20 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F1 Calibration Weight With DAkkS Calibration Certificate > Mass: 20g