Learn More
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
Brand:  Sartorius™ YCW423-00
1109.89 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10727812
  • 1135.00 SEK / Each
Only null left
Estimated Shipment: 18-08-2025
Description
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
  • Without DAkkS Calibration Certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
Plastic
20 g
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight > Plastic; Mass: 20 g