Learn More
Sartorius™ Stainless Steel OIML Class F2 Reference Line Weight
Stainless Steel OIML Class F2 Reference Line Weight without DAkkS Calibration Certificate.
Brand:  Sartorius™ YCW624-00
5980.68 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10103891
  • 6120.00 SEK / Each
Only null left
Estimated Shipment: 18-07-2025
Description
  • Without DAkkS Calibration Certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
Wood
2 kg
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F2 Reference Line Weight > Wood; Mass: 2 kg