Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN110plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand: Memmert™ SN110plusB02073
1509.36 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
SN110plus Sterilizer | |
Drip Tray | |
Rim: 15 mm | |
Stainless Steel |
Product Content Correction
Product Title
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN110plus, 15 mm > For Use With: SN110plus
Spot an opportunity for improvement?Share a Content Correction