Learn More
Macherey-Nagel™ Standard Filter Papers for Qualitative Analysis, MN 617 we, Folded
Manufactured from the same raw materials as ashless papers and are particularly well suited for general laboratory filtrations.
Brand: Macherey-Nagel™ 535012
This item is not returnable.
View return policy
Description
- Folded
- Manufactured from the same raw materials as the ashless grades
- Particularly suited for general laboratory filtrations
- Average ash content is about 0.1%
- The amount of α-cellulose is about 95%
Specifications
85 g/m2 | |
Folded | |
MN 617 we | |
0.22 mm |
Extra soft, smooth surface | |
5 sec. | |
100/Pk. | |
125 mm |
Product Content Correction
Product Title
Macherey-Nagel™ Standard Filter Papers for Qualitative Analysis, MN 617 we, Folded > 125 mm diameter
Spot an opportunity for improvement?Share a Content Correction