Learn More
Standard Precision Buffer Solution pH 1.677, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
167725B1 | Certificate of Analysis | 20/05/2025 | 101677 |
167723H1 | Certificate of Analysis | 03/10/2023 | 101677 |
167723D1 | Certificate of Analysis | 05/07/2023 | 101677 |
167722M1 | Certificate of Analysis | 01/06/2023 | 101677 |
167722F1 | Certificate of Analysis | 22/11/2022 | 101677 |
Product Content Correction
Product Title
Standard Precision Buffer Solution pH 1.677, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction