Learn More
Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 65 mm Shelf Height
Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.
875.00 SEK - 1270.00 SEK
Specifications
Includes | Locking rod |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 65 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15240904
|
Tenak™
TE27202 |
Microtiterplates (10) |
1220.00 SEK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
15250904
|
Tenak™
TE27204 |
Microtiterplates (11) |
1270.00 SEK
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
15200904
|
Tenak™
TE27194 |
Microtiterplates (6) |
990.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15210904
|
Tenak™
TE27196 |
Microtiterplates (7) |
1050.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15220904
|
Tenak™
TE27198 |
Microtiterplates (8) |
1105.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15230904
|
Tenak™
TE27200 |
Microtiterplates (9) |
1160.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15280894
|
Tenak™
TE27190 |
Microtiterplates (4) |
875.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15290894
|
Tenak™
TE27192 |
Microtiterplates (5) |
935.00 SEK
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks come with a locking rod to secure the cryoboxes
- Constructed of stainless steel
Specifications
Locking rod | |
Stainless Steel | |
Chest Frseezers | |
65 mm |