missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 45 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27424
Additional Details : Weight : 2.30000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
5 x 4 | |
Stainless Steel | |
360 x 135 x 233 mm |
Microtiterplates (20) | |
Upright Freezers | |
45 mm |