Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 45 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27548
Additional Details : Weight : 4.20000kg
 View more versions of this product
Product Code. 15250964
  • 2400.00 SEK / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications
Specifications
7 x 6
Stainless Steel
540 x 135 x 325 mm
Microtiterplates (42)
Upright Freezers
45 mm