Learn More
Tenak™ Standard Rack for Upright Freezer, 85 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE23242
Additional Details : Weight : 2.45000kg
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
3 x 4 | |
Stainless Steel | |
560 x 139 x 264 mm |
Boxes (12) | |
Upright Freezers | |
85 mm (Box) |