Learn More
Tenak™ Standard Rack for Upright Freezer, 85 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
1160.00 SEK - 1485.00 SEK
Specifications
Material Stainless Steel
For Use With (Equipment) Upright Freezers
Height (Metric) 85 mm (Box)
View More Specs
Products 7
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
15200714
View Documents Promotion Details
Tenak™
TE23232
Boxes (9)
1200.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
15210714
View Documents Promotion Details
Tenak™
TE23240
Boxes (8)
1200.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
15220714
View Documents Promotion Details
Tenak™
TE23242
Boxes (12)
1230.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
15230714
View Documents Promotion Details
Tenak™
TE23244
Boxes (16)
1340.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
15240714
View Documents Promotion Details
Tenak™
TE23250
Boxes (10)
1340.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
15250714
View Documents Promotion Details
Tenak™
TE23252
Boxes (15)
1485.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
15290704
View Documents Promotion Details
Tenak™
TE23230
Boxes (6)
1160.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Specifications
Stainless Steel
Upright Freezers
85 mm (Box)