Learn More
Tenak™ Standard Rack for Upright Freezer, 85 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE23252
Additional Details : Weight : 2.80000kg
 View more versions of this product
Product Code. 15250714
  • 1485.00 SEK / Each
Estimated Shipment: 21-08-2024
to see stock.
Add to basket
Description
Description
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Specifications
3 x 5
Stainless Steel
683 x 139 x 264 mm
Boxes (15)
Upright Freezers
85 mm (Box)