Learn More
Starch (from Wheat), Millipore™
Specifications
Specifications
| CAS | 9005-25-8 |
| Molecular Formula | (C6H10O5)n |
| MDL Number | MFCD00082026 |
| Quantity | 1 kg |
| Synonym | Starch (from wheat), starch native from wheat, Starch (from wheat) |
| Grade | Research |