missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARS/ABRA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22673-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as an activator of serum response factor (SRF)-dependent transcription possibly by inducing nuclear translocation of MKL1 or MKL2 and through a mechanism requiring Rho-actin signaling.Specifications
STARS/ABRA | |
Polyclonal | |
Unconjugated | |
ABRA | |
ABRA, STARS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
137735, 223513 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BUZ1, Q8N0Z2 | |
ABRA | |
STARS/ABRA Fusion Protein Ag18456 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STARS/ABRA Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction