Learn More
STAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16674-1-AP-150UL
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human STAT2. The calculated molecular weight of STAT2 is 98 kDa, but phosphorylated STAT2 is about 100-113 kDa.
CD95 (Fas, APO-1), a 46 kDa transmembrane glycoprotein, is a cell death receptor of the TNFR superfamily. Stimulation of CD95 results in aggregation of its intracellular death domains, formation of the death-inducing signaling complex (DISC) and activation of caspases. In type I cells caspase 3 is activated by high amounts of caspase 8 generated at the DISC, in type II cells low concentration of caspase 8 activates pathway leading to the release of cytochrome c from mitochondria and activation of caspase 3 by cytochom c. Besides its roles in induction of apoptosis, Fas also triggers pro-inflammatory cytokine responses.Specifications
STAT2 | |
Polyclonal | |
Unconjugated | |
STAT2 | |
ISGF 3, P113, STAT113, STAT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20847, 288774, 6773 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52630, Q9WVL2 | |
Stat2 | |
STAT2 Fusion Protein Ag10168 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.