missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | STAT2 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896365
|
Proteintech
51075-2-AP-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886365
|
Proteintech
51075-2-AP-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a rabbit polyclonal antibody raised against a peptide mapping within human STAT2. The calculated molecular weight of STAT2 is 98 kDa, but phosphorylated STAT2 is about 100-113 kDa.
CD95 (Fas, APO-1), a 46 kDa transmembrane glycoprotein, is a cell death receptor of the TNFR superfamily. Stimulation of CD95 results in aggregation of its intracellular death domains, formation of the death-inducing signaling complex (DISC) and activation of caspases. In type I cells caspase 3 is activated by high amounts of caspase 8 generated at the DISC, in type II cells low concentration of caspase 8 activates pathway leading to the release of cytochrome c from mitochondria and activation of caspase 3 by cytochom c. Besides its roles in induction of apoptosis, Fas also triggers pro-inflammatory cytokine responses.Specifications
STAT2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P52630 | |
6773 | |
Peptide | |
Primary | |
-20°C | |
STAT2 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ISGF 3, P113, STAT113, STAT2 | |
STAT2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STAT2 Rabbit anti-Human, Polyclonal, Proteintech