missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11944-1-AP-20UL
This item is not returnable.
View return policy
Description
The six-transmembrane epithelial antigen of prostate 4 (STEAP4) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 2 and STEAP3), STEAP4 promotes both iron and copper reduction. STEAP4 is highly expressed in placenta, lung, heart and prostate tissues, and overexpressed in prostate cancer cells compared to normal prostate cells. Overexpression of STEAP4 in prostate cells significantly increases cell growth and colony formation, suggesting STEAP4 may have a role in cell proliferation and prostate cancer progression.Specifications
STEAP4 | |
Polyclonal | |
Unconjugated | |
STEAP4 | |
Metalloreductase STEAP4, STAMP2, STEAP family member 4, STEAP4, TIARP, TNFAIP9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117167, 499991, 79689 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V8K1, Q687X5, Q923B6 | |
Steap4 | |
STEAP4 Fusion Protein Ag2545 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STEAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction