Learn More
RETSCH Steel Grinding Sets
Brand:  RETSCH 01.462.0002
25070.50 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 5.20000kg
 View more versions of this product
Product Code. 11994342
  • 26390.00 SEK / Each
Estimated Shipment: 28-08-2024
to see stock.
Specifications
Grinding Set
For grinding without heavy-metal contamination
Steel
50 mL
For RS 200/RS 100 Steel 1,1