Learn More
RETSCH Steel Grinding Sets
26390.00 SEK - 33310.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11994342
|
RETSCH
01.462.0002 |
50 mL |
26390.00 SEK
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11702325
|
RETSCH
01.462.0266 |
250 mL |
33310.00 SEK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
Specifications
Grinding Set | |
Steel |