Learn More
Stellasterol, TRC
Product Code. 17968984
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17968984 25 mg 25mg
17949084 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17968984 Supplier Toronto Research Chemicals Supplier No. TRCS68668025MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2465-11-4
Molecular Formula C28 H46 O
Molecular Weight (g/mol) 398.66
Synonym (3beta,5alpha,22E)-Ergosta-7,22-dien-3-ol, 5alpha-Ergosta-7,22-dien-3beta-ol, (22E)-Ergosta-7,22-dien-3beta-ol, (22E,24R)-Ergosta-7,22-dien-3beta-ol, (24S)-24-Methyl-5alpha-cholesta-7,22-dien-3beta-ol, 24-Methyl-5alpha-cholesta-7,22-dien-3beta-ol, 5-Dihydroergosterin, Ergosta-7,22-dienol, Stellasterin, alpha-Dihydroergosterol;
IUPAC Name (3S,5S,9R,10S,13R,14R,17R)-10,13-dimethyl-17-[(E,1R,4R)-1,4,5-trimethylhex-2-enyl]-2,3,4,5,6,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES C[C@@]12[C@](CC[C@]2([H])[C@H](C)/C=C/[C@H](C)C(C)C)([H])C3=CC[C@](C[C@H](CC4)O)([H])[C@@]4(C)[C@@]3([H])CC1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C28H46O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-8,10,18-22,24-26,29H,9,11-17H2,1-6H3/b8-7+/t19-,20+,21-,22-,24+,25-,26-,27-,28+/m0/s1
Formula Weight 398.3549
Chemical Name or Material Stellasterol