Learn More
Sampling Systems™ SteriWare™ Forceps (Tweezers)
Our Sterile single use SteriWare™ Forceps (Tweezers) are completely manufactured in a cleanroom to ensure cleanliness. They come with full documentation that confirms food grade and sterility.
Brand: Sampling Systems™ 8630A-115
Description
- Our disposable forceps are available in both sterile and non sterile versions.
- They are suitable for lifting and moving small objects such as tablets, samples etc.
- The handles have undulations moulded into them to give you extra grip.
- These lightweight tweezers are suitable for food, chemical, pharmaceutical, or cosmetic applications.
- These tweezers can be used straight from the bag.
- The sterile tweezers have been processed by gamma irradiation.
- Made from food grade PS.
- Conforms to the latest FDA and EU requirements.
- BSE/TSE free
- To avoid contamination, they have been manufactured and then packed in a class 7 cleanroom.
- They come with full documentation that confirms food grade and sterility.
Specifications
Polystyrene | |
115 mm | |
Non-sterile | |
Bag |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
070588 | Certificate of Compliance | 08/12/2022 | 8630A-115 |