missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STIM1 Mouse anti-Human, Mouse, Rat, Clone: 1D3A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66189-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a type 1 transmembrane protein that mediates Ca2+ influx after depletion of intracellular Ca2+ stores by gating of store-operated Ca2+ influx channels (SOCs). It is one of several genes located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocrotical carcinoma, and lung, ovarian, and breast cancer. This gene may play a role in maligcies and disease that involve this region, as well as early hematopoiesis, by mediating attachment to stromal cells. This gene is oriented in a head-to-tail configuration with the ribonucleotide reductase 1 gene (RRM1), with the 3′ end of this gene situated 1.6 kb from the 5′ end of the RRM1 gene.Specifications
STIM1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70302, P84903, Q13586 | |
Mouse | |
Protein A | |
RUO | |
20866, 361618, 6786 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
1D3A3 | |
Unconjugated | |
STIM1 | |
Stim1 | |
STIM1 Fusion Protein Ag17914 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STIM1 Mouse anti-Human, Mouse, Rat, Clone: 1D3A3, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction