Learn More
STIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15218-1-AP-150UL
This item is not returnable.
View return policy
Description
STIP1 is an adaptor protein that coordinates the functions of HSP70 and HSP90 in protein folding. It is thought to assist in the transfer of proteins from HSP70 to HSP90 by binding both HSP90 and substrate-bound HSP70. STIP1 also stimulates the ATPase activity of HSP70 and inhibits the ATPase activity of HSP90, suggesting that it regulates both the conformations and ATPase cycles of these chaperones (Song and Masison, 2005 ).Specifications
STIP1 | |
Polyclonal | |
Unconjugated | |
STIP1 | |
HOP, Hsc70/Hsp90 organizing protein, IEF SSP 3521, P60, STI1, STI1L, STIP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10963, 192277, 20867 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35814, P31948, Q60864 | |
Stip1 | |
STIP1 Fusion Protein Ag7388 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
STIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction