missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STK17A (DRAK1) Recombinant Human Protein

STK17A (DRAK1) is a serine/threonine kinase.

Brand:  Invitrogen™ PV4061

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10717953

  • 79000.00 SEK / 1mg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

STK17A (DRAK1) is localized in the nucleus and is involved in apoptosis.

Specifications

Specifications

NP_004751
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
STK17A (DRAK1)
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
74kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
DAPK Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers