missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK17A (DRAK1) Recombinant Human Protein
STK17A (DRAK1) is a serine/threonine kinase.
18240.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_004751 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 74kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498503
|
Invitrogen™
PR7034B |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10717953
|
Invitrogen™
PV4061 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
STK17A (DRAK1) is localized in the nucleus and is involved in apoptosis.
Specifications
NP_004751 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
DAPK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74kDa | |
RUO - research use only | |
STK17A (DRAK1) | |
Human | |
GST Tag | |
Frozen |