missing translation for 'onlineSavingsMsg'
Learn More
Invitrogen™ Human STK23 (MSSK1), GST Tag Recombinant Protein Product Code.: 11573607

Invitrogen™ Human STK23 (MSSK1), GST Tag Recombinant Protein

Product Code. p-7180594
10 μg, 10µg
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 11573607

Brand: Invitrogen™ PV3880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

STK23 (MSSK1), also known as SRPK3, is a muscle-specific member of the SRPKs. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UPE1
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 26576
Molecular Weight (g/mol) 86.5 kDa
Name Human STK23 (MSSK1), GST Tag
Quantity 10 μg
Gene Alias MSSK1; MSSK-1; muscle-specific serine kinase 1; muscle-specific serine kinase 1; MSSK1; serine arginine rich protein-specific kinase 3; serine/arginine-rich protein specific kinase 3; serine/arginine-rich protein-specific kinase 3; serine/threonine kinase 23; serine/threonine kinase 23, muscle-specific serine kinase 1 70; serine/threonine-protein kinase 23; serine/threonine-protein kinase SRPK3; SFRS protein kinase 3; Srpk3; SR-protein-specific kinase 3; SRSF protein kinase 3; STK23
Common Name SRPK3
KinaseFamily SRPK Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody)
Kinase Group Ser/Thr Kinases: CMGC Group
Shipping Condition Dry Ice
System Compatibility Zin-LYTE™
Tag Position N-Terminal
Target Entry STK23
Target Gene SFRS protein kinase 3
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human STK23 (MSSK1), GST Tag Recombinant Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.