Learn More
Wesbart™ Storage Bin
A useful way of keeping loose items safe within a freezer
Brand: Wesbart™ BIN827
Additional Details : Weight : 0.80000kg
Description
Specifications
Storage Bin | |
Freezer | |
Handle on One End and a Label Holder | |
280 mm |
Anodized Aluminum, Stainless Steel | |
Keeping Loose Items Safe within a Freezer | |
280 mm | |
0.8 kg |